Bromotrimethylsilane, 98%
Bromotrimethylsilane, 98%
Bromotrimethylsilane, 98%
Thermo Scientific Chemicals

Bromotrimethylsilane, 98%

CAS: 2857-97-8 | C3H9BrSi | 153.1 g/mol
数量:
5 g
25 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 186650250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2857-97-8
Titration Argentometric>=97.5 %
Appearance (Color)Clear colorless to yellow
Refractive index1.4200 to 1.4250 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Silylation reagent
RUO – Research Use Only