Bromotrimetilsilano, 98 %, Thermo Scientific Chemicals
Bromotrimetilsilano, 98 %, Thermo Scientific Chemicals
Bromotrimetilsilano, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bromotrimetilsilano, 98 %, Thermo Scientific Chemicals

CAS: 2857-97-8 | C3H9BrSi | 153.1 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
25 g
100 g
500 g
Envase:
Frasco de vidrio
Número de catálogo 186651000
Precio (EUR)
116,00
Each
Cantidad:
100 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
116,00
Each
Identificadores químicos
CAS2857-97-8
EspecificacionesSpecification SheetHoja de especificaciones
Titration Argentometric>=97.5 %
Appearance (Color)Clear colorless to yellow
Refractive index1.4200 to 1.4250 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reactivo de formación de enlaces C-X, reactivo de sililación
RUO – Research Use Only