3,4-Dimethoxyphenol, 97%
3,4-Dimethoxyphenol, 97%
3,4-Dimethoxyphenol, 97%
Thermo Scientific Chemicals

3,4-Dimethoxyphenol, 97%

CAS: 2033-89-8 | C8H10O3 | 154.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
View all
Glass Bottle
Catalog number 186700500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS2033-89-8
IUPAC Name3,4-dimethoxyphenol
Molecular FormulaC8H10O3
InChI KeySMFFZOQLHYIRDA-UHFFFAOYSA-N
SMILESCOC1=CC=C(O)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light beige
Melting point77°C to 82°C
GC>=96.0 %
Appearance (Form)Crystalline powder
AppearanceMay darken on storage
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only