O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%
Thermo Scientific Chemicals

O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%

O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, C7H4F5NO.HCl, CAS Number-57981-02-9 | CAS: 57981-02-9 | C7H5ClF5NO | 249.57 g/mol
製品番号(カタログ番号) 186710010
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS57981-02-9
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Crystalline powder, crystals, flakes and/or chunks
Titration Argentometric97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only