O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%
Thermo Scientific Chemicals

O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%

O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, C7H4F5NO.HCl, CAS Number-57981-02-9 | CAS: 57981-02-9 | C7H5ClF5NO | 249.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 186712500
Price (USD)
68.00
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
68.00
Each
Chemical Identifiers
CAS57981-02-9
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Crystalline powder, crystals, flakes and/or chunks
Titration Argentometric97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only