m-Phenetidine, 98%
m-Phenetidine, 98%
m-Phenetidine, 98%
Thermo Scientific Chemicals

m-Phenetidine, 98%

CAS: 621-33-0 | C8H11NO | 137.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 186720050
Price (USD)
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS621-33-0
IUPAC Name3-ethoxyaniline
Molecular FormulaC8H11NO
InChI KeyWEZAHYDFZNTGKE-UHFFFAOYSA-N
SMILESCCOC1=CC=CC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear brown
Infrared spectrumConforms
Refractive index1.5650 to 1.5670 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only