CEDIA™ Opiate Drugs of Abuse Controls
Product Image
Thermo Scientific™

CEDIA™ Opiate Drugs of Abuse Controls

Accurately detect opiates in patient samples with Thermo Scientific™ CEDIA™ Opiate Drugs of Abuse Assays, for use on a wide array of clinical chemistry analyzers.
Have Questions?
Catalog NumberQuantity
18689341 x 15 mL
Catalog number 1868934
Price (USD)
-
Request A Quote
Quantity:
1 x 15 mL

Accurately detect opiates in patient samples with Thermo Scientific™ CEDIA™ Opiate Drugs of Abuse Assays, for use on a wide array of clinical chemistry analyzers.

The scope of drugs of abuse testing is expanding rapidly, and Thermo Scientific CEDIA assays continue to meet the challenge. CEDIA immunoassays are valued worldwide for their accuracy, precision, lot-to-lot dependability and stable shelf life. We offer the most extensive menu for drugs of abuse testing and we are continuing to develop new assays to respond to ever-emerging drug screening needs.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionCEDIA Opiate 2K High Control
Quantity1 x 15 mL
TypeControl
Detectable AnalytesOpiate
Sample TypeUrine
Unit SizeEach