Petri Dish
Buehler™
Petri Dish
For use with Spectrophotometer CM-5
Have Questions?
Catalog NumberType
1870-712Optical Glass
Catalog number 1870-712
Price (USD)
-
Type:
Optical Glass
When used with Spectrohotometer CM-5, this Petri Dish enables the reflectance of opaque paste, powders, grains and granular materials to be measured.
  • Optical glass
  • 45 x 15 mm
Specifications
Dimensions (L x W)45 x 15 mm
For Use With (Application)Enable The Reflectance Measurements of Opaque Paste, Powders, Grains and Granular Materials
For Use With (Equipment)Spectrophotometer CM-5
ModelCM-A128
TypeOptical Glass
Unit SizeEach