4-Amino-1-benzylpiperidine, 98%
4-Amino-1-benzylpiperidine, 98%
4-Amino-1-benzylpiperidine, 98%
Thermo Scientific Chemicals

4-Amino-1-benzylpiperidine, 98%

CAS: 50541-93-0 | C12H18N2 | 190.29 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 187660250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS50541-93-0
IUPAC Name1-benzylpiperidin-4-amine
Molecular FormulaC12H18N2
InChI KeyYUBDLZGUSSWQSS-UHFFFAOYSA-N
SMILESNC1CCN(CC2=CC=CC=C2)CC1
さらに表示
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with HClO4>=97.5 %
GC>=97.5 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only