2-Thiopheneacetic acid, 98%
2-Thiopheneacetic acid, 98%
2-Thiopheneacetic acid, 98%
Thermo Scientific Chemicals

2-Thiopheneacetic acid, 98%

CAS: 1918-77-0 | C6H6O2S | 142.17 g/mol
数量:
5 g
25 g
100 g
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 187731000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1918-77-0
IUPAC Name2-(thiophen-2-yl)acetic acid
Molecular FormulaC6H6O2S
InChI KeySMJRBWINMFUUDS-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=CS1
さらに表示
Appearance (Color)White to grey to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point61°C to 66°C
Titration with NaOH97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only