4-Bromothioanisole, 97%
4-Bromothioanisole, 97%
4-Bromothioanisole, 97%
Thermo Scientific Chemicals

4-Bromothioanisole, 97%

CAS: 104-95-0 | C7H7BrS | 203.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 188191000
Price (USD)
222.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
222.00
Each
Chemical Identifiers
CAS104-95-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Appearance (Form)low melting crystalline mass
GC>=96.0 %
Appearance (Form)Crystalline powder or crystals and/or chunks or
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only