2,6-Difluoroaniline, 97%
2,6-Difluoroaniline, 97%
2,6-Difluoroaniline, 97%
Acros Organics

2,6-Difluoroaniline, 97%

CAS: 5509-65-9 | C6H5F2N | 129.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 188271000
also known as 18827
HSN 29214290
Price (INR)
61,227.00
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
61,227.00
Each
Chemical Identifiers
CAS5509-65-9
IUPAC Name2,6-difluoroaniline
Molecular FormulaC6H5F2N
InChI KeyODUZJBKKYBQIBX-UHFFFAOYSA-N
SMILESNC1=C(F)C=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown
Appearance (Form)Liquid
AppearanceMay darken on storage
Infrared spectrumConforms
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only