3-(Trifluoromethyl)benzaldehyde, 97%
3-(Trifluoromethyl)benzaldehyde, 97%
3-(Trifluoromethyl)benzaldehyde, 97%
Thermo Scientific Chemicals

3-(Trifluoromethyl)benzaldehyde, 97%

CAS: 454-89-7 | C8H5F3O | 174.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 188440250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS454-89-7
IUPAC Name3-(trifluoromethyl)benzaldehyde
Molecular FormulaC8H5F3O
InChI KeyNMTUHPSKJJYGML-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light orange
Infrared spectrumConforms
Refractive index1.4639 to 1.4659 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only