Methyllithium, 1.6M sol. in diethyl ether (± 5% w/v), AcroSeal™, Thermo Scientific Chemicals
Methyllithium, 1.6M sol. in diethyl ether (± 5% w/v), AcroSeal™, Thermo Scientific Chemicals
Methyllithium, 1.6M sol. in diethyl ether (± 5% w/v), AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyllithium, 1.6M sol. in diethyl ether (± 5% w/v), AcroSeal™, Thermo Scientific Chemicals

製品番号(カタログ番号)数量パッケージ
188758000800mLAcroSeal™ Glass Bottle
188751000100mLAcroSeal™ Glass bottle
製品番号(カタログ番号) 188758000
価格(JPY)
-
数量:
800mL
パッケージ:
AcroSeal™ Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS917-54-4
IUPAC Namemethyllithium
Molecular FormulaCH3Li
InChI KeyDVSDBMFJEQPWNO-UHFFFAOYSA-N
SMILES[Li]C
さらに表示
Appearance (Color)Clear to hazy colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only