1,2,4-Triazole, sodium salt, 90%, technical
1,2,4-Triazole, sodium salt, 90%, technical
1,2,4-Triazole, sodium salt, 90%, technical
Thermo Scientific Chemicals

1,2,4-Triazole, sodium salt, 90%, technical

CAS: 41253-21-8 | C2H2N3Na | 91.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 188990100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS41253-21-8
IUPAC Namesodium 4H-1,2,4-triazol-4-ide
Molecular FormulaC2H2N3Na
InChI KeyNVMNEWNGLGACBB-UHFFFAOYSA-N
SMILES[Na+].[N-]1C=NN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to brown
Titration with HClO4>=88 %
Identification(sodium): passes test
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only