3-(3,4,5-Trimethoxyphenyl)propionic acid, 98%
3-(3,4,5-Trimethoxyphenyl)propionic acid, 98%
3-(3,4,5-Trimethoxyphenyl)propionic acid, 98%
Thermo Scientific Chemicals

3-(3,4,5-Trimethoxyphenyl)propionic acid, 98%

CAS: 25173-72-2 | C12H16O5 | 240.25 g/mol
Have Questions?
Catalog NumberQuantity
1891600505 g
Catalog number 189160050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS25173-72-2
IUPAC Name3-(3,4,5-trimethoxyphenyl)propanoic acid
Molecular FormulaC12H16O5
InChI KeyZCYXGVJUZBKJAI-UHFFFAOYSA-N
SMILESCOC1=CC(CCC(O)=O)=CC(OC)=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point100°C to 105°C
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only