Sodium borohydride, 98+%, powder
Sodium borohydride, 98+%, powder
Sodium borohydride, 98+%, powder
Thermo Scientific Chemicals

Sodium borohydride, 98+%, powder

CAS: 16940-66-2 | BH4Na | 37.83 g/mol
数量:
5 g
10 g
100 g
500 g
2.5 kg
パッケージ:
Plastic Bottle
製品番号(カタログ番号) 189305000
価格(JPY)
-
数量:
500 g
パッケージ:
Plastic Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS16940-66-2
IUPAC Namesodium boranuide
Molecular FormulaBH4Na
InChI KeyYOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES[BH4-].[Na+]
さらに表示
Appearance (Form)Powder
Appearance (Color)White
Titration Iodometric>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only