Trypan Blue
Trypan Blue
Trypan Blue
Thermo Scientific Chemicals

Trypan Blue

CAS: 72-57-1 | C34H24N6Na4O14S4 | 960.79 g/mol
Have Questions?
Catalog NumberQuantityPackaging
189355000500 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 189355000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS72-57-1
IUPAC Nametetrasodium (3Z)-5-amino-3-[2-(4'-{2-[(2Z)-8-amino-1-oxo-3,6-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Molecular FormulaC34H24N6Na4O14S4
InChI KeyWMYVHJWZUUEZNE-ARWFNKCKSA-J
SMILES[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1\C(=O)C2=C(N)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2\C(=O)C3=C(N)C=C(C=C3C=C2S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Dye content>=60 %
UV spectrumComplies
Appearance (Form)Powder
Appearance (Color)Dark blue to brown-black
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only