Patent Blue VF, pure, indicator grade
Patent Blue VF, pure, indicator grade
Patent Blue VF, pure, indicator grade
Thermo Scientific Chemicals

Patent Blue VF, pure, indicator grade

CAS: 129-17-9 | C27H31N2NaO6S2 | 566.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 189411000
Price (EUR)
250,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
250,00
Each
Chemical Identifiers
CAS129-17-9
IUPAC Namesodium bis[4-(diethylamino)phenyl](2,4-disulfonatophenyl)methylium
Molecular FormulaC27H31N2NaO6S2
InChI KeySJEYSFABYSGQBG-UHFFFAOYSA-M
SMILES[Na+].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1S([O-])(=O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
UV spectrumComplies
Appearance (Color)Dark blue-green to dark purple
Suitability(as acid-base indicator) Passes test
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only