Tempshield™ Cryo Gloves - Shoulder Length
Thermo Scientific™
Tempshield™ Cryo Gloves - Shoulder Length
Multi-layered construction allows for thermal protection, flexibility, and dexterity which are essential features when function is important and safety is critical. High performance, thermal inner lining wicks moisture away from hands, maintaining comfort over extended periods.
Catalog NumberSize
189449Large
189448Medium
189450X-Large
Catalog number 189449
Price (GBP)
-
Size:
Large
Multi-layered construction allows for thermal protection, flexibility, and dexterity which are essential features when function is important and safety is critical. High performance, thermal inner lining wicks moisture away from hands, maintaining comfort over extended periods. Safety certified to meet EU standards for CE Cat. III: EN511, EN388, EN ISO 21420. Available in wrist, elbow, mid-arm, shoulder-length options.
  • Provides splash protection
  • Comfortable, lightweight, breathable
Specifications
Certifications/ComplianceCE Cat. III: EN 511, EN 388, EN ISO 21420
DescriptionShoulder Length
For Use With (Application)Cryogenic Preservation
Length (English)28 in.
Length (Metric)71 cm
Product TypeWaterproof Cryogenic Gloves
SizeLarge
Unit SizeEach