Nigrosine, pure, water soluble, high purity biological stain
Nigrosine, pure, water soluble, high purity biological stain
Nigrosine, pure, water soluble, high purity biological stain
Thermo Scientific Chemicals

Nigrosine, pure, water soluble, high purity biological stain

CAS: 8005-03-6 | C22H14N6Na2O9S2 | 616.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 189481000
Price (USD)
67.65
Special offer
Online exclusive
Ends: 28-Dec-2026
79.00
Save 11.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
67.65
Special offer
Online exclusive
Ends: 28-Dec-2026
79.00
Save 11.35
Each
Chemical Identifiers
CAS8005-03-6
IUPAC Namedisodium (3E)-5-amino-6-[(1E)-2-(4-nitrophenyl)diazen-1-yl]-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
Molecular FormulaC22H14N6Na2O9S2
InChI KeyHKBVRFLHNUEVRO-HMDKGIQMSA-L
SMILES[Na+].[Na+].NC1=C2C(=O)\C(=N/NC3=CC=CC=C3)C(=CC2=CC(=C1\N=N\C1=CC=C(C=C1)[N+]([O-])=O)S([O-])(=O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(0.1 % in water) Opaque black solution
Infrared spectrumConforms
UV spectrumComplies
UV Lambda max570 to 580 nm (c=0.02 g/L, 50 % ethanol)
Appearance (Color)Black
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only