Phenylpyruvic acid, sodium salt monohydrate, 98%
Phenylpyruvic acid, sodium salt monohydrate, 98%
Phenylpyruvic acid, sodium salt monohydrate, 98%
Thermo Scientific Chemicals

Phenylpyruvic acid, sodium salt monohydrate, 98%

CAS: 122049-54-1 | C9H7O3 | 163.15 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 189730250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS122049-54-1
Appearance (Form)Powder
Water7 to 12 % (K.F.)
Infrared spectrumConforms
Titration with HClO497.5 to 102.5 %
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only