Thermo Scientific™
Square Cover Slips
 Related applications:
Anatomical Pathology
Utilize Thermo Scientific™ Square Cover Slips, which are manufactured with colorless borosilicate glass, for a wide variety of microscopy applications. Our cover slips feature high-spectral transmission quality and a finely adaptive refractive index suited for microscopy. Each cover slip is individually selected for uniform quality in thickness, flatness and size. These cover slips are made for use in anatomical pathology, hematology, microbiology, urinalysis and research.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Virtually colorless appearance
  • Very low levels of inclusions, striae, bubbles, and streaks
  • High-spectral transmission
  • Finely adaptive refractive index suited for microscopes
  • Extremely flat surface
Documents
Manuals & protocols