2-Bromo-2',4'-dimethoxyacetophenone, 98%
2-Bromo-2',4'-dimethoxyacetophenone, 98%
2-Bromo-2',4'-dimethoxyacetophenone, 98%
Thermo Scientific Chemicals

2-Bromo-2',4'-dimethoxyacetophenone, 98%

CAS: 60965-26-6 | C10H11BrO3 | 259.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1900100101 gGlass bottle
Catalog number 190010010
Price (EUR)
70,65
Online exclusive
78,20
Save 7,55 (10%)
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
70,65
Online exclusive
78,20
Save 7,55 (10%)
Each
Chemical Identifiers
CAS60965-26-6
IUPAC Name2-bromo-1-(2,4-dimethoxyphenyl)ethan-1-one
Molecular FormulaC10H11BrO3
InChI KeyPKVBZABQCCQHLD-UHFFFAOYSA-N
SMILESCOC1=CC=C(C(=O)CBr)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Infrared spectrumConforms
Melting point101°C to 105°C
GC>=97.5 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only