Chrome Azurol S
Chrome Azurol S
Chrome Azurol S
Thermo Scientific Chemicals

Chrome Azurol S

CAS: 1667-99-8 | C23H13Cl2Na3O9S | 605.277 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 190050250
Price (EUR)
90,65
Online exclusive
100,00
Save 9,35 (9%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
90,65
Online exclusive
100,00
Save 9,35 (9%)
Each
Chemical Identifiers
CAS1667-99-8
IUPAC Nametrisodium 5-{[(1E)-3-carboxylato-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene](2,6-dichloro-3-sulfonatophenyl)methyl}-2-hydroxy-3-methylbenzoate
Molecular FormulaC23H13Cl2Na3O9S
InChI KeyFUIZKNBTOOKONL-DPSBJRLESA-K
SMILES[Na+].[Na+].[Na+].CC1=CC(=CC(C([O-])=O)=C1O)C(=C1\C=C(C)C(=O)C(=C1)C([O-])=O)\C1=C(Cl)C=CC(=C1Cl)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
UV Lambda max424 to 436 nm (c=0.03 g/l in H2O)
Appearance (Color)Dark brown to black
Infrared spectrumConforms
UV spectrumComplies
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only