Product Image
Thermo Scientific™
Solid Door Option for Large Chamber
Protection for your light sensitive samples
Have Questions?
Catalog NumberdimVoltage
1900665TS
Catalog number 1900665TS
Price (MXN)
-
Solid outer door for Thermo Scientific™ chamber - replaces standard outer glass door to protect samples against light. Recommended for light sensitive applications. Factory-installed option, please order in conjunction with environmental chamber. F
  • Replaces standard outer glass door to protect samples against light
  • Recommended for light sensitive applications
  • Factory-installed option, please order in conjunction with environmental chamber
  • Choose based on size of chamber: small (for 311 L / 11 cu. ft. models) or large (for 821 L / 29 cu.ft. models)
  • Optional inner glass door enables visual check of samples with no major disturbance of temperature / humidity levels
Specifications
DescriptionSolid door for large chamber, replaces standard outer glass door, factory installed
Door StyleSolid door for large chamber
For Use With (Application)light sensitive applications
For Use With (Equipment)order with chamber models 3920, 3940, 3948, 3960, 3962
IncludesFactory installed solid door
LightingLED
TypeSolid Door Option
Unit SizeEach