Solid Door Option with Inner Glass Door for Large Chamber
Product Image
Thermo Scientific™
Solid Door Option with Inner Glass Door for Large Chamber
Protection for your light sensitive samples
Have Questions?
Catalog NumberDoor Style
1900667TSSolid with Inner Glass
Catalog number 1900667TS
Price (MXN)
-
Door Style:
Solid with Inner Glass
Solid outer door for Thermo Scientific™ chamber - replaces standard outer glass door to protect samples against light. Recommended for light sensitive applications. Factory-installed option, please order in conjunction with environmental chamber. F
  • Replaces standard outer glass door to protect samples against light
  • Recommended for light sensitive applications
  • Factory-installed option, please order in conjunction with environmental chamber
  • Choose based on size of chamber: small (for 311 L / 11 cu. ft. models) or large (for 821 L / 29 cu.ft. models)
  • Optional inner glass door enables visual check of samples with no major disturbance of temperature / humidity levels
Specifications
DescriptionSolid door with inner glass door for large chamber, replaces standard outer glass door, factory installed
Door StyleSolid with Inner Glass
For Use With (Application)Incubation, light sensitive applications
For Use With (Equipment)Chamber Models 3920, 3940, 3948, 3960, 3962
IncludesFactory installed solid door with inner glass door
LightingLED
TypeSolid Door Option
Unit SizeEach