This product will be discontinued on May 1, 2017. Please see notification below.

CaptureSelect™ IgG-Fc (Hu) Affinity Matrix

Catalog number: 190082210

 Related applications: Antibody Purification | Chromatography | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CaptureSelect™ IgG-Fc (Hu) Affinity Matrix can be used to purify all four subclasses of human IgG by high affinity binding to the Fc region of IgGs. Unlike protein A, it is selective for human IgG and binds to human IgG3 as well. This affinity matrix does not cross-react with bovine IgG (FCS) and can be used for purification of:

• IVIG from plasma and serum
• Monoclonal IgG subclasses 1-4 from cell cultures
• Monoclonal and polyclonal IgG subclasses 1-4 from transgenic sources
• Recombinant human Fc fusion proteins (both full length Fc as well as CH2 deleted)
• No cross-binding to bovine IgG (FCS)

Discontinuation May 1, 2017

Date: October 2016.

Dear Customer,

Please be informed that we intend to discontinue this product line on May 1st 2017.

We have been able to improve stability and elution characteristics of the affinity matrix. The improved product, CaptureSelect™ FcXL Affinity Matrix, has been commercially available for over two years and will replace the original product IgG-Fc (Hu) and we plan to discontinue the current product after a period of 6 months on May 1, 2017.

Please find the improved version item codes:
SKU Description Size Size Type
194328005 CAPTURESELECT FCXL AFF MX 5 mL
194328010 CAPTURESELECT FCXL AFF MX 10 mL
194328050 CAPTURESELECT FCXL AFF MX 50 mL

In addition, the FcXL affinity matrix is available in 1- and 5- mL pre-packed column format, and resin slurry is available at large scale and also suitable for bioprocess applications. For this product a ligand leakage ELISA and a regulatory support file is available.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Column Type: Bulk Resin
Format: Bulk Media
Immobilized Phase: Anti IgG-Fc
Product Line: CaptureSelect™
Product Size: 10 mL

Contents & storage

Store at 2–8°C.

Documents

Manuals & protocols