CaptureSelect™ IgG-Fc (Hu) Affinity Matrix

Catalog number: 190082250

 Related applications: Antibody Purification | Chromatography | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CaptureSelect™ IgG-Fc (Hu) Affinity Matrix can be used to purify all four subclasses of human IgG by high affinity binding to the Fc region of IgGs. Unlike protein A, it is selective for human IgG and binds to human IgG3 as well. This affinity matrix does not cross-react with bovine IgG (FCS) and can be used for purification of:

• IVIG from plasma and serum
• Monoclonal IgG subclasses 1-4 from cell cultures
• Monoclonal and polyclonal IgG subclasses 1-4 from transgenic sources
• Recombinant human Fc fusion proteins (both full length Fc as well as CH2 deleted)
• No cross-binding to bovine IgG (FCS)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: Bulk Media
Column Type: Bulk Resin
Product Size: 50 mL
Immobilized Phase: Anti IgG-Fc

Contents & storage

Store at 2–8°C.

Documents

Manuals & protocols