New Coccine, pure
New Coccine, pure
New Coccine, pure
Thermo Scientific Chemicals

New Coccine, pure

CAS: 2611-82-7 | C20H11N2Na3O10S3 | 604.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 190231000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2611-82-7
IUPAC Nametrisodium (8Z)-7-oxo-8-[2-(4-sulfonatonaphthalen-1-yl)hydrazin-1-ylidene]-7,8-dihydronaphthalene-1,3-disulfonate
Molecular FormulaC20H11N2Na3O10S3
InChI KeyIVNZBWNBYXERPK-DZGBHZPSSA-K
SMILES[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=C4C=CC=CC4=C(C=C3)S([O-])(=O)=O)C(=O)C=C2)C(=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<0.2 % (in water)
Appearance (Color)Red to brown
UV spectrumComplies
Dye content>=75 %
Solubility(0.1 % in water) Clear red to very dark red
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only