CaptureSelect™ Human Growth Hormone Affinity Matrix

Catalog number: 190316010

 Related applications: Chromatography | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This is one of a group of CaptureSelect™ protein purification affinity resins developed for the simple, single-step purification of a variety human plasma and serum proteins. This unique selection of affinity purification resins can purify proteins or other non-antibody biomolecules from either recombinant or native sources with a high degree of affinity and specificity, and are immobilized on a high capacity agarose support.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Column Type: Bulk Resin
Format: Bulk Media
Immobilized Phase: Anti Human Growth Hormone
Product Line: CaptureSelect™
Product Size: 10 mL

Contents & storage

Store at 2–8°C.

Documents

Manuals & protocols