CaptureSelect™ Antithrombin III Affinity Matrix
Thermo Scientific™

CaptureSelect™ Antithrombin III Affinity Matrix

This is one of a group of Thermo Scientific™ CaptureSelect™ protein purification affinity resins developed for the simple, single-step purification of a variety human plasma and serum proteins.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
19031701010 mL
1903170055 mL
19031705050 mL
Catalog number 190317010
Price (USD)
2,300.00
Each
Quantity:
10 mL
Price (USD)
2,300.00
Each

This is one of a group of CaptureSelect protein purification affinity resins developed for the simple, single-step purification of a variety human plasma and serum proteins. This unique selection of affinity purification resins can purify proteins or other non-antibody biomolecules from either recombinant or native sources with a high degree of affinity and specificity, and are immobilized on a high capacity agarose support.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity10 mL
Stationary PhaseAnti Antithrombin III
Capacity (Metric)>6 mg/mL matrix
FormResin Slurry
Particle Size90 μm
Product LineCaptureSelect
TypeBulk Resin
Unit SizeEach
Contents & Storage
• Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light