Thermo Scientific Chemicals

Lithium aluminum hydride, 95%, powder, Thermo Scientific Chemicals

產品號碼: 190320250
25 g, Each
Thermo Scientific Chemicals

Lithium aluminum hydride, 95%, powder, Thermo Scientific Chemicals

產品號碼: 190320250
25 g, Each
Quantity
產品號碼: 190320250
價格 (TWD)

化學識別

CAS
16853-85-3
IUPAC Name
lithium(1+) alumanuide
Molecular Formula
AlH4Li
InChI Key
OCZDCIYGECBNKL-UHFFFAOYSA-N
SMILES
[Li+].[AlH4-]
Appearance (Color)
White to light grey
Assay
>=94 %
Appearance (Form)
Crystalline powder

說明

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text