Thermo Scientific Chemicals

Lithium aluminum hydride, 95%, powder, Thermo Scientific Chemicals

Catalog number: 190325000
500 g, Each
Thermo Scientific Chemicals

Lithium aluminum hydride, 95%, powder, Thermo Scientific Chemicals

Catalog number: 190325000
500 g, Each
Quantity

Chemical Identifiers

CAS
16853-85-3
IUPAC Name
lithium(1+) alumanuide
Molecular Formula
AlH4Li
InChI Key
OCZDCIYGECBNKL-UHFFFAOYSA-N
SMILES
[Li+].[AlH4-]
Appearance (Color)
White to light grey
Assay
>=94 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text