Aluminum oxide, for chromatography, acidic, Brockmann I, 50-200 μm, 90A
Aluminum oxide, for chromatography, acidic, Brockmann I, 50-200 μm, 90A
Thermo Scientific Chemicals

Aluminum oxide, for chromatography, acidic, Brockmann I, 50-200 μm, 90A

 
Have Questions?
Catalog NumberQuantity
1904000101 kg
Catalog number 190400010
Price (EUR)
137,00
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
137,00
Each
Add to cart
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle size=<5 % (> 200 um)
Appearance (Form)Crystalline powder
pH4.2 to 4.8
Particle size=<10 % (< 50 um)
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only