Thermo Scientific Chemicals

Tris(triphenylphosphine)rhodium(I) chloride, 98%, Thermo Scientific Chemicals

Catalog number: 190420050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Tris(triphenylphosphine)rhodium(I) chloride, 98%, Thermo Scientific Chemicals

Catalog number: 190420050
5 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
14694-95-2
IUPAC Name
λ¹-rhodium(1+) tris(triphenylphosphane) chloride
Molecular Formula
C54H45ClP3Rh
InChI Key
IXAYKDDZKIZSPV-UHFFFAOYSA-M
SMILES
[Cl-].[Rh+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Red-brown to brown
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Gravimetry
10.8 to 11.3 % (Rh)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text