You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Home
▸
Shop All Products
▸
Incubators
▸
Incubator Accessories
▸
Data Output, 4 to 20 milliamp
Thermo Scientific™
Data Output, 4 to 20 milliamp
For Use With:
Environmental Chamber (3962)
Have Questions?
Catalog Number
For Use With (Equipment)
190512
Environmental Chamber (3962)
Catalog number
190512
Price (BRL)
-
Request A Quote
For Use With (Equipment):
Environmental Chamber (3962)
Integrate Thermo Scientific™ Data Output Accessories for your environmental chamber or incubator to increase functionality and compatibility.
Specifications
Description
4-20 milliamp Data Output
For Use With (Equipment)
Environmental Chamber (3962)
Installation
Factory Installed
Lighting
LED
Type
Data Output
Unit Size
Each