iConnect™ Split/Splitless with Backflush (SSL-BKF) Injector Module for TRACE 1300 Series GC
iConnect™ Split/Splitless with Backflush (SSL-BKF) Injector Module for TRACE 1300 Series GC
iConnect™ Split/Splitless with Backflush (SSL-BKF) Injector Module for TRACE 1300 Series GC
iConnect™ Split/Splitless with Backflush (SSL-BKF) Injector Module for TRACE 1300 Series GC
Thermo Scientific™
iConnect™ Split/Splitless with Backflush (SSL-BKF) Injector Module for TRACE 1300 Series GC
Save analysis time and gain robustness against heavy contamination with the iConnect Split/Splitless with Integrated Backflush (SSL-BKF).
Catalog NumberInjection Mode
19070011Split/Splitless
Catalog number 19070011
Price (JPY)
-
Injection Mode:
Split/Splitless
Extend the life of your analytical column and maximize laboratory throughput by eliminating high boiling compounds. The Thermo Scientific iConnect SSL-BKF injector module enables you to easily add pre-column backflush to your split or splitless injection.
Thanks to the self-adjusting pressure during backflushing, the integrated pneumatic control eliminates complexity and the cost of an additional auxiliary gas module, and facilitates method setup.
  • Suitable for all capillary columns (50 μm to 530 μm inner diameter)
  • Supports concurrent solvent recondensation (CSR) for large volume injections (up to 50 μL)
  • Supports P&T/TD/HS by special adapter
  • Compatible with the Merlin Microseal™ septum
  • Maximum temperature: 400°C
SSL injector with integrated concurrent backflush
  • Reverse the flow inside the injector
  • Eliminate heavy or “undesired” compounds concurrently during the analysis run
  • Increase throughput
SSL injector advantages
  • Perform reproducible, accurate, and robust sample injection
  • Reduce bleed and extend septa lifetime
  • Implement existing and validated methods quickly and easily
  • Use many standard consumables such as liners, septa, and ferrules already in your lab
  • Gain easy and immediate access to the septum and liner for simple and quick maintenance without tools or removing gas lines thanks to TRACE 1300 Series iConnect modules
  • Extract and restore the injector body quickly for cleaning when needed, such as after difficult, sticky samples
Reduce downtime, simplify maintenance, and enhance GC performance
iConnect injectors and detectors redefine GC and GC-MS flexibility and usability. In two minutes, without training or tools, you can change these modules to:
  • Reconfigure your system for a specific workflow or method
  • Perform maintenance and dramatically reduce maintenance downtime
  • Switch injectors and detectors between TRACE GC systems
Specifications
DescriptioniConnect Split/Splitless (SSL) with Backflush (BF) Injector module
QuantityEach
For Use With (Equipment)TRACE 1300 Series Gas Chromatographs; AI/AS 1310, AI/AS 1610, TriPlus 100 LS, TriPlus RSH, TriPlus RSH SMART, and TriPlus 500 HS Autosamplers
Injection ModeSplit/Splitless
Product TypeiConnect SSL Injector with Backflush
Unit SizeEach