iConnect™ Pulsed Discharge Detector (PDD) for TRACE™ 1300 and 1600 Series GC
Thermo Scientific™

iConnect™ Pulsed Discharge Detector (PDD) for TRACE™ 1300 and 1600 Series GC

Use the iConnect™ Pulsed Discharge Detector (PDD) for excellent selectivity and sensitivity for gas detection on TRACE™ 1300 and 1600 Series GC systems.

Have Questions?
Catalog NumberDetection Mode
19070014Pulsed Discharge
Catalog number 19070014
Price (USD)
-
Detection Mode:
Pulsed Discharge

Achieve excellent selectivity and sensitivity for gas detection using the Thermo Scientific™ iConnect™ Pulsed Discharge Detector (PDD) for TRACE™ 1300 and 1600 Series gas chromatographs. This versatile, self-sufficient GC detector and the electronic and pneumatic circuits necessary for control and gas supply can be installed on any TRACE 1300 or 1600 Series GC, regardless of its previous configuration, allowing for GC system modularity.

Switch Detectors in Two Minutes
iConnect injectors and detectors redefine flexibility and usability in GC and GC-MS analysis. In two minutes, without special training or tools, you can swap out these modules and reconfigure your instrument for a specific workflow or method. This unique capability can also enable continuous operation or dramatically reduce maintenance downtime -- just replace dirty units with clean backups and be back to running samples again in minutes.

Options for Expansion
For budget-conscious labs, iConnect modules offer the flexibility to start with a single-channel instrument, and expand to multi-injector or multi-detector configurations to accommodate new applications or increased throughput requirements.

The iConnect Pulsed Discharge Detector offers selectivity and sensitivity for critical PDD analyses.

  • Maximum Temperature: 400°C
  • Linear Dynamic Range: 105
  • MDL: low pg range
Specifications
DescriptioniConnect Pulsed Discharge Detector (PDD) module
Detection ModePulsed Discharge
Dynamic RangeMinimum Detection: Low pg Range
For Use With (Equipment)TRACE 1300 and 1600 Series Gas Chromatographs
LinearityLinear Range: 105
Detector TypePDD
Product TypeiConnect Pulsed Discharge Detector
Temperature RangeOperating Temperature: 400°C
Unit Size-