iConnect™ Programmable Temperature Vaporizing (PTV) Injector Module
iConnect™ Programmable Temperature Vaporizing (PTV) Injector Module
iConnect™ Programmable Temperature Vaporizing (PTV) Injector Module
Thermo Scientific™
iConnect™ Programmable Temperature Vaporizing (PTV) Injector Module
Providing fast heating and cooling performance, iConnect™ PTV Injectors are ideal for trace analysis of dirty matrices and thermally labile compounds.
Have Questions?
Catalog NumberQuantity
19070020Each
Catalog number 19070020
Price (USD)
8,790.00
Each
Quantity:
Each
Price (USD)
8,790.00
Each
Gain greater versatility in gas chromatography with instant connect (iConnect™) injectors for Thermo Scientific™ TRACE™ 1300 and 1600 Series GC systems. The Thermo Scientific™ iConnect Programmable Temperature Vaporizing (PTV) Injector Module combines the discrimination-free performance of a cold injector with the robustness of vaporizing injectors. This PTV injector offers fast heating and cooling performance, making it ideal for trace analysis of dirty matrices and thermally labile compounds. Its unique design and multi-mode operations preserve sample integrity in all situations. The injector features fast cooling, a cryogenic option for initial sub-ambient temperatures, a backflush option, and easy maintenance.
FOR US ONLINE SALES ONLY
  • Supports hot/cold split and splitless modes as well as large-volume (solvent split) and on-column (TPOC) injections
  • Compatible with the Merlin Microseal™ septum
  • Temperature range with air cooling: ambient +5°C up to 450°C
  • Cryogenic option minimum temperature: -100°C with liquid nitrogen; -50°C with CO2
  • Temperature programming of up to 3 ramps at up to 870°C/min
Fast cooling
  • Compact and limited thermal-mass design and efficient forced air circulation system
  • A convenient cryogenic option is available for initial sub-ambient temperature applications
Quick and trouble-free maintenance
  • Remove the liner from the top and gain complete access to the injector chamber
PTV injector with integrated concurrent backflush option
  • Reverse the flow inside the injector
  • Eliminate heavy or “undesired” compounds concurrently during analysis
  • Increase throughput
Syringe-autosampler compatibility
  • Syringe 365D0391 is required for correct operation with Thermo Scientific™ TriPlus 100 LS and TriPlus RSH SMART Autosamplers
Reduce downtime, simplify maintenance, and enhance GC performance
iConnect injectors and detectors for TRACE 1300 and 1600 Series GC redefine GC and GC-MS flexibility and usability. In two minutes, without training or tools, you can change these modules to:
  • Reconfigure your system for a specific workflow or method
  • Perform maintenance and dramatically reduce maintenance downtime
  • Switch injectors and detectors between any TRACE GC Series systems
Specifications
DescriptioniConnect Programmable Temperature Vaporizing (PTV) Injector module
QuantityEach
For Use With (Equipment)TRACE 1300 and 1600 Series Gas Chromatographs; AI/AS 1310, AI/AS 1610, TriPlus 100 LS, TriPlus RSH, and TriPlus RSH SMART Autosamplers
Injection ModeHot/Cold Split, Splitless, Large-Volume (Solvent Split), and On Column (TPOC) Injections
Product TypeiConnect PTV Injector
Unit SizeEach