iConnect™ Programmable Temperature Vaporizing with Backflush (PTV-BKF) Injector Module for TRACE 1300 Series GC
iConnect™ Programmable Temperature Vaporizing with Backflush (PTV-BKF) Injector Module for TRACE 1300 Series GC
iConnect™ Programmable Temperature Vaporizing with Backflush (PTV-BKF) Injector Module for TRACE 1300 Series GC
iConnect™ Programmable Temperature Vaporizing with Backflush (PTV-BKF) Injector Module for TRACE 1300 Series GC
Thermo Scientific™
iConnect™ Programmable Temperature Vaporizing with Backflush (PTV-BKF) Injector Module for TRACE 1300 Series GC
Save analysis time and gain robustness against heavy contamination. Experience the simplicity of self-regulated backflush injections.
Have Questions?
Catalog NumberQuantity
19070021Each
Catalog number 19070021
Price (USD)
-
Quantity:
Each
Extend the life of your analytical column, maximize laboratory throughput, and increase the robustness of your GC system. To eliminate unwanted high boilers, the pre-column backflush configuration combines backflush capabilities for normal and large volume injections.
The Thermo Scientific™ iConnect™ PTV-BKF injector removes the complexity and cost of an additional auxiliary gas module and facilitates method setup thanks to the self-adjusting pressure during backflushing.
  • Supports hot/cold and splitless modes as well as large-volume (solvent split)
  • Compatible with the Merlin Microseal™ septum
  • Temperature range with air cooling: ambient +5°C up to 450°C
  • Cryogenic option minimum temperature: -100°C with liquid nitrogen; -50°C with CO2
  • Temperature programming of up to 3 ramps at up to 870°C/min
Fast cooling
  • Compact and limited thermal-mass design and efficient forced air circulation system
  • A convenient cryogenic option is available for initial sub-ambient temperature applications
Quick and trouble-free maintenance
  • Remove the liner from the top and gain complete access to the injector chamber
PTV injector with integrated concurrent backflush option
  • Reverse the flow inside the injector
  • Eliminate heavy or “undesired” compounds concurrently during analysis
  • Increase throughput
Reduce downtime, simplify maintenance, and enhance GC performance
iConnect injectors and detectors for TRACE 1300 Series GC redefine GC and GC-MS flexibility and usability. In two minutes, without training or tools, you can change these modules to:
  • Reconfigure your system for a specific workflow or method
  • Perform maintenance and dramatically reduce maintenance downtime
  • Switch injectors and detectors between any TRACE GC Series systems
Specifications
DescriptioniConnect Programmable Temperature Vaporizing (PTV) with Backflush (BF) Injector module
QuantityEach
For Use With (Equipment)TRACE 1300 Series Gas Chromatographs; AI/AS 1310, AI/AS 1610, TriPlus 100 LS, TriPlus RSH, and TriPlus RSH SMART Autosamplers
Injection ModeHot/Cold Split, Splitless, Large-Volume (Solvent Split)
Product TypeiConnect PTV Injector with Backflush
Unit SizeEach