SuperScript V Reverse Transcriptase, 40,000 units - FAQs
SuperScript V Reverse Transcriptase, 40,000 units - FAQs
View additional product information for SuperScript V Reverse Transcriptase - FAQs (19090050, 19090200, 19090010)
This product does not have any FAQs available.