Ethyl cellulose, ethoxyl content 48%, 10 cps
Ethyl cellulose, ethoxyl content 48%, 10 cps
Ethyl cellulose, ethoxyl content 48%, 10 cps
Thermo Scientific Chemicals

Ethyl cellulose, ethoxyl content 48%, 10 cps

CAS: 9004-57-3 | C23H24N6O4 | 448.483 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
Packaging:
Glass bottle
Catalog number 190962500
Price (USD)
122.00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
122.00
Each
Chemical Identifiers
CAS9004-57-3
IUPAC Name2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate
Molecular FormulaC23H24N6O4
InChI KeyARSKJXYLLONUAJ-UHFFFAOYSA-N
SMILESCCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Viscosity8 to 12 cP (5% in 80/20 toluene/ethanol, 25°C)
Assay47.5 to 49.5 % (Ethoxyl content)
Appearance (Form)Powder or granules
Appearance (Color)White to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only