Cesium acetate, 97%, pure
Cesium acetate, 97%, pure
Cesium acetate, 97%, pure
Thermo Scientific Chemicals

Cesium acetate, 97%, pure

CAS: 3396-11-0 | C2H3CsO2 | 191.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 191041000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3396-11-0
IUPAC Namecaesium(1+) acetate
Molecular FormulaC2H3CsO2
InChI KeyZOAIGCHJWKDIPJ-UHFFFAOYSA-M
SMILES[Cs+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder and/or chunks
Titration with HClO496.0 to 104.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only