Code Reader™ 2600 Barcode Readers
Fisher Scientific™
Code Reader™ 2600 Barcode Readers
Reads all 1D and 2D barcode symbologies
Have Questions?
카탈로그 번호포함유형
19104470
FISHERMIS-19104470-EACH으로도 사용됨
Bluetooth, BatteryBarcode Reader Handle Kit
카탈로그 번호 19104470
FISHERMIS-19104470-EACH으로도 사용됨
제품 가격(KRW)
-
포함:
Bluetooth, Battery
유형:
Barcode Reader Handle Kit
Reads all 1D and 2D barcode symbologies
  • Easy to use, lightweight, and high performing
  • Features durable, rechargeable battery plus programmable buttons for custom workflow
  • Battery status LED indicators with fuel gauge
  • User feedback with vibration, audible tones and LED
  • Patented dual field optics, both high density and wide field in same unit
  • High speed, omnidirectional reading of 1D and 2D barcodes
  • Unequaled performance on rounded, curved and shiny surfaces
  • Multiple programmable buttons for customized work flow processes
  • Easy to clean, disinfectant ready, IP65 housing for healthcare use
  • Powerful data management capability with JavaScript
  • Available in Handle and Palm kits
사양
포함Bluetooth, Battery
유형Barcode Reader Handle Kit
Unit SizeEach