CaptureSelect™ MultiSpecies Albumin Depletion Product
Thermo Scientific™

CaptureSelect™ MultiSpecies Albumin Depletion Product

This is one of a group of Thermo Scientific™ CaptureSelect™ Proteomics Toolbox™ affinity resins which are unique affinity resins developed for the depletion of proteins from human plasma or serum samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1910853055 mL
19108531010 mL
19108535050 mL
Catalog number 191085305
Price (USD)
1,250.00
Each
Quantity:
5 mL
Price (USD)
1,250.00
Each

This is one of a group of CaptureSelect Proteomics Toolbox™ affinity resins which are unique affinity resins developed for the depletion of proteins from human plasma or serum samples. These affinity ligands are immobilized on a high capacity agarose support for easy, lower-pressure protein depletion or sample prep applications.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity5 mL
Stationary PhaseAnti-albumin
Capacity (Metric)15 - 25 mg albumin/mL resin
FormResin Slurry
Particle Size35 μm
Product LineCaptureSelect
TypeAlbumin Depletion Product
Unit SizeEach
Contents & Storage
• Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light

Frequently asked questions (FAQs)

Can I use the High Select Top14 Abundant Protein Depletion Midi Spin Columns to deplete proteins from rodent samples?

The High Select Top14 Abundant Protein Depletion Midi Spin Columns have only been validated for human samples. We do not recommend using them for protein depletion in rodent samples. For protein depletion in rodent samples, we recommend the following multi-species depletion resins:

  • CaptureSelect MultiSpecies Albumin Depletion Product (Cat. No. 191085305)
  • Pierce Albumin Serum Depletion Kits (Cat. No. 89875)


  • Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.