N-Benzylbenzamide, 99%
N-Benzylbenzamide, 99%
N-Benzylbenzamide, 99%
Thermo Scientific Chemicals

N-Benzylbenzamide, 99%

CAS: 1485-70-7 | C14H13NO | 211.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
View all
Glass bottle
Catalog number 191170050
Price (USD)
32.65
Online exclusive
36.30
Save 3.65
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
32.65
Online exclusive
36.30
Save 3.65
Each
Chemical Identifiers
CAS1485-70-7
IUPAC NameN-benzylbenzamide
Molecular FormulaC14H13NO
InChI KeyLKQUCICFTHBFAL-UHFFFAOYSA-N
SMILESO=C(NCC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point104°C to 107°C
Appearance (Color)White
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only