Notice: This product will be discontinued on March 20, 2021.
Thermo Scientific™

CaptureSelect™ IgG-Fc (ms) Affinity Matrix

Catalog number:  191285550

Related applications:

Antibody Purification

| Chromatography

| Protein Purification & Isolation

Back to top

Description

CaptureSelect™ IgG-Fc (ms) Affinity Matrix has been specifically designed to purify IgG from multiple species by high affinity binding to the Fc region of IgGs . Unlike protein G, gentle elution conditions can be used to recover the product. This affinity matrix can be used for purification of IgG from a broad range of species, such as human, mouse, rat, rabbit, cow, horse, and sheep. A complete list of species can be found in the product information sheet.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Column Type: Bulk Resin
Product Line: CaptureSelect™
Format: Bulk Media
Stationary Phase: Anti IgG-Fc

Contents & storage

Store at 2–8°C.