CaptureSelect™ Human Albumin Affinity Matrix

Catalog number: 19129705L

 Related applications: Antibody Purification | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CaptureSelect™ Human Albumin Affinity Matrix has been specifically designed for the purification of human albumin and albumin fusion proteins.

Features of this affinity matrix include:

• Highly selective for human albumin
• Unique elution characteristics allowing for gentle elution of albumin fusion proteins
• Excellent scalability
• Non-animal-derived

The high selectivity and yields obtained using CaptureSelect™ Human Albumin Affinity Matrix enable a robust and efficient purification process with excellent purity obtained in one step. The mild elution conditions make the resin suitable for the purification of albumin fusion proteins, especially those that are pH sensitive.

Free of Animal Components
Our CaptureSelect™ products are affinity ligands created by a proprietary technology based on Camelid-derived single domain antibody fragments. The ligand is a 13 kDa single domain fragment comprising the 3 CDRs that form the antigen binding domain, efficiently produced by the yeast Saccharomyces cerevisiae in a production process free of animal components.

Main Characteristics:

Matrix: agarose-based, aldehyde-activated
Average particle size: 65 ± 10 µm
Ligand: CaptureSelect™ HSA affinity ligand
Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
Binding capacity: ~15 mg albumin/mL resin
Elution conditions (neutral pH): 20 mM Tris, pH 7.4 with 2 M MgCl2 or with 1 M NaCl + 50%(v/v) propylene glycol or with 1 M NaCl + 0.5 M L-arginine
Elution conditions (acidic pH): 0.1 M glycine, pH 3.0
Flow characteristics: 200–400 cm/h (up to 1 bar)
Formulation buffer: 20%(v/v) ethanol
Caution: For manufacturing, processing, or repacking.

Specifications

Column Type: Bulk Resin
Flow Rate: 200 to 400 cm⁄hr
Format: Bottle(s)
Immobilized Phase: Anti HSA
Particle Size: 65 micron
Product Line: CaptureSelect™
Product Size: 5 L
Support Matrix: Agarose based, aldehyde activated

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols