CaptureSelect™ IgG1 (Hu) Affinity Matrix
Thermo Scientific™

CaptureSelect™ IgG1 (Hu) Affinity Matrix

Thermo Scientific™ CaptureSelect™ IgG1 (Hu) Affinity Matrix can be used for the purification and isolation of IgG1, without cross-reactivity to other immunoglobulins.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
19130301010 mL
1913030055 mL
19130305050 mL
Catalog number 191303010
Price (MXN)
-
Quantity:
10 mL

CaptureSelect IgG1 (Hu) Affinity Matrix can be used for the purification and isolation of IgG1, without cross-reactivity to other immunoglobulins, like other human IgG subclasses, IgA, and IgM, from complex sources like plasma, serum, and cell culture supernatants.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity10 mL
Stationary PhaseAnti-IgG1
Binding Capacity>8 g/L Resin
FormResin Slurry
Particle Size35 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
• Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light